missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluorophenylacetone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6153514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Fluorophenylacetone | |
Yellow | |
47°C | |
C9H9FO | |
MFCD00000325 | |
2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone # | |
CC(=O)CC1=CC=CC=C1F | |
152.168 | |
152.17 | |
Liquid |
2836-82-0 | |
1.077 g/mL | |
85°C (185°F) | |
1.5 | |
25 g | |
BANVZEUCJHUPOI-UHFFFAOYSA-N | |
1-(2-fluorophenyl)propan-2-one | |
76086 | |
98% |
Chemical Identifiers
2836-82-0 | |
152.168 | |
BANVZEUCJHUPOI-UHFFFAOYSA-N | |
76086 | |
CC(=O)CC1=CC=CC=C1F |
C9H9FO | |
MFCD00000325 | |
2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone # | |
1-(2-fluorophenyl)propan-2-one |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 220-627-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only