Please login to your online account to display your discounted pricing

(2-Fluorophenyl)methanesulfonyl chloride, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material (2-Fluorophenyl)methanesulfonyl chloride
CAS 24974-71-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H6ClFO2S
Formula Weight 208.64
MDL Number MFCD04117455
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07770DA

  SDS
fisher scientific company
MO07770DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.64
Quantity 1g
Assay 95%
CAS Min % 95.0
Chemical Name or Material (2-Fluorophenyl)methanesulfonyl chloride
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Assay Percent Range 95%
Molecular Formula C7H6ClFO2S
MDL Number MFCD04117455
CAS 24974-71-8
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.