Please login to your online account to display your discounted pricing

2-Fluorophenyl isocyanate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170320250

 View more versions of this product

Catalog No. AC170320250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 137.11
Quantity 25g
Color Clear colorless to slightly brown
TSCA TSCA
EINECS Number 240-804-0
Density 1.2220g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Fluorophenyl isocyanate
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H4FNO
Linear Formula FC6H4NCO
MDL Number MFCD00001996
CAS 16744-98-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
Flammable liquid and vapo
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5115 to 1.5135 (20°C, 589nm)

Specifications

Chemical Name or Material 2-Fluorophenyl isocyanate
Name Note 97%
CAS 16744-98-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4FNO
Linear Formula FC6H4NCO
Formula Weight 137.11
MDL Number MFCD00001996
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
Flammable liquid and vapo

SDS & Safety Documents