missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorophenol, TCI America™

Manufacturer:  TCI America F0150250G

 View more versions of this product

Catalog No. F0150250G



Specifications

Specifications

2-Fluorophenol
Yellow
367-12-4
MFCD00002155
o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
C1=CC=C(C(=C1)O)F
112.103
112.10
≥98.0% (GC)
152°C
250g
C6H5FO
1993
HFHFGHLXUCOHLN-UHFFFAOYSA-N
2-fluorophenol
9707
Clear Liquid at 20°C

Chemical Identifiers

367-12-4
112.103
HFHFGHLXUCOHLN-UHFFFAOYSA-N
9707
C1=CC=C(C(=C1)O)F
C6H5FO
MFCD00002155
o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
2-fluorophenol
Safety and Handling

Safety and Handling

RTECSNumber : SL4530000

TSCA : Yes

Documents