missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorophenol, 98%

Manufacturer:  Alfa Aesar™ A1497936

 View more versions of this product

Catalog No. AAA1497936



Specifications

Specifications

2-Fluorophenol
Phenol-like
14°C to 16°C
367-12-4
MFCD00002155
1905112
HFHFGHLXUCOHLN-UHFFFAOYSA-N
2-fluorophenol
9707
98%
46°C (114°F)
150°C to 152°C
500g
C6H5FO
UN2924
o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
C1=CC=C(C(=C1)O)F
112.103
112.1

Chemical Identifiers

367-12-4
112.103
HFHFGHLXUCOHLN-UHFFFAOYSA-N
9707
C1=CC=C(C(=C1)O)F
C6H5FO
MFCD00002155
o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
2-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

RTECSNumber : SL4530000

TSCA : TSCA

Documents