missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorophenethyl alcohol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 310970050

 View more versions of this product

Catalog No. AC310970050


 

Specifications Description & Specifications

Specifications

≥98.5%
140.16
Clear colorless
100.0
98.5
Authentic
C8H9FO
MFCD00002887
99%
Liquid
5g
1.04g/mL
1.5080 to 1.5100 (20°C, 589nm)
Glass bottle
50919-06-7
2-Fluorophenethyl alcohol
FC6H4CHCHOH
99%

Chemical Identifiers

50919-06-7
100.0
C8H9FO
140.16
2-Fluorophenethyl alcohol, 99%
98.5
99%
FC6H4CHCHOH
MFCD00002887
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA