missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluorophenacyl bromide, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446670250
Specifications
2-Fluorophenacyl bromide | |
25g | |
97.5% min. (GC) | |
2-bromo-1-2'-fluorophenyl ethanone, 2-bromo-1-2-fluorophenyl ethan-1-one, 2-bromo-1-2-fluorophenyl ethanone, 2-bromo-2'-fluoroacetophenone, 2-fluorophenacyl bromide, 2-fluorophenacylbromide, alpha-bromo-2-fluoroacetophenone, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone | |
C1=CC=C(C(=C1)C(=O)CBr)F | |
217.04 | |
217.04 |
61.0°C to 63.0°C (0.1 Torr) | |
655-15-2 | |
C8H6BrFO | |
QDNWNJSLWKHNTM-UHFFFAOYSA-N | |
2-bromo-1-(2-fluorophenyl)ethanone | |
2737449 | |
98% |
Chemical Identifiers
655-15-2 | |
217.04 | |
2-bromo-1-2'-fluorophenyl ethanone, 2-bromo-1-2-fluorophenyl ethan-1-one, 2-bromo-1-2-fluorophenyl ethanone, 2-bromo-2'-fluoroacetophenone, 2-fluorophenacyl bromide, 2-fluorophenacylbromide, alpha-bromo-2-fluoroacetophenone, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone | |
2-bromo-1-(2-fluorophenyl)ethanone |
C8H6BrFO | |
QDNWNJSLWKHNTM-UHFFFAOYSA-N | |
2737449 | |
C1=CC=C(C(=C1)C(=O)CBr)F |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger