Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoronicotinic acid, 97%

$37.22 - $248.62

Chemical Identifiers

Chemical Name or Material 2-Fluoronicotinic acid
CAS 393-55-5
Molecular Formula C6H4FNO2
Formula Weight 141.1
MDL Number MFCD00040744
Beilstein 3612
Synonym 2-Fluoropyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1944203

  SDS
alfa aesar™
L1944203
1g Each for $90.72

AAL1944206

  SDS
alfa aesar™
L1944206
5g Each for $248.62

AAL19442MD

  SDS
alfa aesar™
L19442MD
250mg Each for $37.22
Description & Specifications

Specifications

Formula Weight 141.1
Chemical Name or Material 2-Fluoronicotinic acid
MDL Number MFCD00040744
CAS 393-55-5
Synonym 2-Fluoropyridine-3-carboxylic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C6H4FNO2
Beilstein 3612
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈170°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.