Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoromandelic acid, 99%

$53.49 - $53.49

Chemical Identifiers

Chemical Name or Material 2-Fluoromandelic acid
CAS 389-31-1
Molecular Formula C8H7FO3
Formula Weight 170.14
MDL Number MFCD03095382
Synonym 2-Fluoro-alpha-hydroxyphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2568903

  SDS
alfa aesar™
B2568903
1g Each for $53.49

AAB2568909

  SDS
alfa aesar™
B2568909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Fluoromandelic acid
Melting Point 113° to 116°C
Quantity 1g
CAS 389-31-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Molecular Formula C8H7FO3
Formula Weight 170.14
MDL Number MFCD03095382
Synonym 2-Fluoro-alpha-hydroxyphenylacetic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.