Please login to your online account to display your discounted pricing

2-Fluoroethanol, 95%, Acros Organics


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC170830250

 View more versions of this product

Catalog No. AC170830250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 2-Fluoroethanol
Quantity 25g
CAS 371-62-0
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C2H5FO
Formula Weight 64.06
Synonym Ethylene fluorohydrin

Specifications

Chemical Name or Material 2-Fluoroethanol
Assay Percent Range 95%
Molecular Formula C2H5FO
Formula Weight 64.06
Synonym Ethylene fluorohydrin