missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoroethanol, 95%, Acros Organics

Manufacturer:  Acros Organics AC170830050

 View more versions of this product

Catalog No. AC170830050



Specifications

Specifications

2-Fluoroethanol
34°C
100.0°C to 104.0°C
5g
95%
FCH2CH2OH
GGDYAKVUZMZKRV-UHFFFAOYSA-N
2-fluoroethanol
9737
95%
1.1080g/mL
Glass bottle
-26.5°C
371-62-0
C2H5FO
ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
C(CF)O
64.059
64.06

Chemical Identifiers

371-62-0
64.059
ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
2-fluoroethanol
C2H5FO
GGDYAKVUZMZKRV-UHFFFAOYSA-N
9737
C(CF)O
Documents