Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoroethanol, 95%

Chemical Identifiers

Chemical Name or Material 2-Fluoroethanol
CAS 371-62-0
Molecular Formula FCH2CH2OH
Formula Weight 64.06
MDL Number MFCD00002828
UN Number UN3384
Beilstein 1730857
Synonym Ethylene fluorhydrin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0173503

  SDS
alfa aesar™
L0173503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0173506

  SDS
alfa aesar™
L0173506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 64.06
Chemical Name or Material 2-Fluoroethanol
CAS 371-62-0
MDL Number MFCD00002828
Molecular Formula FCH2CH2OH
Assay 95%
Health Hazard 2 GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Synonym Ethylene fluorhydrin
Flash Point 31°C (87°F)
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 1 Danger
Melting Point −27°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
TSCA TSCA
UN Number UN3384
RTECS Number KL1575000
Boiling Point 102° to 104°C
Beilstein 1730857

Applications
2-Fluoroethanol, is used in the microwave spectra of two isotopic species of 2-fluoroethanol have been investigated. The Gg conformer of 2-fluoroethanol is isolated in low-temperature noble gas matrices. Single-photon IR photolysis of 2-fluoroethanol in solid argon has been investigated.

Solubility
Fully miscible.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.