Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorocyclohexanone, 96%

$99.14 - $265.19

Chemical Identifiers

Chemical Name or Material 2-Fluorocyclohexanone
CAS 694-82-6
Molecular Formula C6H9FO
Formula Weight 116.13
MDL Number MFCD01076476
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603203

  SDS
alfa aesar™
H2603203
1g Each for $265.19

AAH26032MD

  SDS
alfa aesar™
H26032MD
250mg Each for $99.14
Description & Specifications

Specifications

Formula Weight 116.13
Quantity 1g
Molecular Formula C6H9FO
Recommended Storage −30° to −10°C
Assay 96%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 78°C (17mmHg)
MDL Number MFCD01076476
CAS 694-82-6
Flash Point 71°C (160°F)
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Fluorocyclohexanone
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C