missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluorocinnamic acid, predominantly trans, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1530314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Fluorocinnamic acid | |
451-69-4 | |
Odorless | |
MFCD00004370 | |
1863963 | |
IOUDZAFBPDDAMK-AATRIKPKSA-N | |
(E)-3-(2-fluorophenyl)prop-2-enoic acid | |
735833 | |
≥98% |
predominantly trans | |
179°C to 183°C | |
C9H7FO2 | |
25 g | |
2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid | |
OC(=O)\C=C\C1=CC=CC=C1F | |
166.15 | |
166.15 |
Chemical Identifiers
451-69-4 | |
166.15 | |
IOUDZAFBPDDAMK-AATRIKPKSA-N | |
735833 | |
OC(=O)\C=C\C1=CC=CC=C1F |
C9H7FO2 | |
MFCD00004370 | |
2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid | |
(E)-3-(2-fluorophenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-195-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only