missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorobiphenyl, 98%

Manufacturer:  Alfa Aesar™ A1222714

 View more versions of this product

Catalog No. AAA1222714



Description

Description

Applications
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications

Specifications

2-Fluorobiphenyl
72°C to 76°C
321-60-8
MFCD00000317
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2F
172.202
172.2
248°C
25g
C12H9F
2043175
KLECYOQFQXJYBC-UHFFFAOYSA-N
1-fluoro-2-phenylbenzene
67579
98%

Chemical Identifiers

321-60-8
172.202
KLECYOQFQXJYBC-UHFFFAOYSA-N
67579
C1=CC=C(C=C1)C2=CC=CC=C2F
C12H9F
MFCD00000317
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene
1-fluoro-2-phenylbenzene
Safety and Handling

Safety and Handling

RTECSNumber : DV5291000

TSCA : TSCA

SDS Documents