Please login to your online account to display your discounted pricing

2-Fluorobiphenyl 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluorobiphenyl
Name Note 97%
CAS 321-60-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H9F
Linear Formula C6H5C6H4F
Formula Weight 172.2
MDL Number MFCD00000317
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146840010

  SDS
thermo fisher scientific
AC146840010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC146840050

  SDS
thermo fisher scientific
AC146840050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.2
Quantity 1g
EINECS Number 206-290-7
TSCA TSCA
RTECS Number DV5291000
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Fluorobiphenyl
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H9F
Linear Formula C6H5C6H4F
MDL Number MFCD00000317
CAS 321-60-8
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.