Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorobiphenyl-4-boronic acid, 97%

$198.57 - $198.57

Chemical Identifiers

Chemical Name or Material 2-Fluorobiphenyl-4-boronic acid
CAS 178305-99-2
Molecular Formula C12H10BFO2
Formula Weight 216.02
MDL Number MFCD01075707
Synonym 3-Fluoro-4-phenylbenzeneboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1563406

  SDS
alfa aesar™
L1563406
5g Each for $198.57
Description & Specifications

Specifications

Formula Weight 216.02
Chemical Name or Material 2-Fluorobiphenyl-4-boronic acid
Assay 97%
Molecular Formula C12H10BFO2
MDL Number MFCD01075707
CAS 178305-99-2
Synonym 3-Fluoro-4-phenylbenzeneboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈242° to 250°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.