missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorobenzylamine, 97%

Manufacturer:  Alfa Aesar™ A1562306

 View more versions of this product

Catalog No. AAA1562306



Specifications

Specifications

2-Fluorobenzylamine
Amine-like
5g
C7H8FN
UN2735
2-fluorobenzylamine, 2-fluorophenyl methanamine, benzenemethanamine, 2-fluoro, o-fluorobenzylamine, benzylamine, o-fluoro, 1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorophenyl methylamine, o-fluorobenzyl amine
C1=CC=C(C(=C1)CN)F
125.146
125.15
67°C (152°F)
80°C to 82°C (18mmHg)
89-99-6
MFCD00008107
508109
LRFWYBZWRQWZIM-UHFFFAOYSA-N
(2-fluorophenyl)methanamine
66649
97%

Chemical Identifiers

89-99-6
125.146
LRFWYBZWRQWZIM-UHFFFAOYSA-N
66649
C1=CC=C(C(=C1)CN)F
C7H8FN
MFCD00008107
2-fluorobenzylamine, 2-fluorophenyl methanamine, benzenemethanamine, 2-fluoro, o-fluorobenzylamine, benzylamine, o-fluoro, 1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorophenyl methylamine, o-fluorobenzyl amine
(2-fluorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : TSCA

Documents