missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzylamine, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 171940100

 View more versions of this product

Catalog No. AC171940100



Description

Description

Specifications

Specifications

2-Fluorobenzylamine
67°C
Glass bottle
1.09
Colorless to Yellow
89-99-6
100.0
C7H8FN
MFCD00008107
LRFWYBZWRQWZIM-UHFFFAOYSA-N
(2-fluorophenyl)methanamine
66649
Liquid
1.0900g/mL
Authentic
1.5160 to 1.5190
73.0°C to 75.0°C (13.0mmHg)
10g
95.0
95% min. (GC)
FC6H4CH2NH2
1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorobenzylamine, 2-fluorophenyl methanamine, 2-fluorophenyl methylamine, benzenemethanamine, 2-fluoro, benzylamine, o-fluoro, o-fluorobenzyl amine, o-fluorobenzylamine
C1=CC=C(C(=C1)CN)F
125.15
125.15
96%

Chemical Identifiers

89-99-6
125.15
LRFWYBZWRQWZIM-UHFFFAOYSA-N
66649
C1=CC=C(C(=C1)CN)F
C7H8FN
MFCD00008107
1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorobenzylamine, 2-fluorophenyl methanamine, 2-fluorophenyl methylamine, benzenemethanamine, 2-fluoro, benzylamine, o-fluoro, o-fluorobenzyl amine, o-fluorobenzylamine
(2-fluorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 201-957-9

SDS Documents