Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorobenzyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material 2-Fluorobenzyl chloride
CAS 345-35-7
Molecular Formula C7H6ClF
Formula Weight 144.58
MDL Number MFCD00000892
UN Number UN2920
Beilstein 471699
Synonym alpha-Chloro-2-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1156614

  SDS
alfa aesar™
A1156614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1156622

  SDS
alfa aesar™
A1156622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.58
Chemical Name or Material 2-Fluorobenzyl chloride
TSCA TSCA
Assay ≥98%
Molecular Formula C7H6ClF
MDL Number MFCD00000892
CAS 345-35-7
UN Number UN2920
Beilstein 471699
Synonym alpha-Chloro-2-fluorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
Flash Point 57°C (134°F)
Odor Pungent
Boiling Point 175°C
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.