Learn More
2-Fluorobenzyl chloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1156622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Fluorobenzyl chloride | |
1.216 | |
57°C (134°F) | |
C7H6ClF | |
MFCD00000892 | |
UN2920 | |
2-fluorobenzyl chloride, 1-chloromethyl-2-fluorobenzene, o-fluorobenzyl chloride, alpha-chloro-o-fluorotoluene, benzene, 1-chloromethyl-2-fluoro, alpha-chloro-2-fluorotoluene, 2-fluorobenzylchloride, 1-chloromethyl-2-fluoro-benzene, toluene, alpha-chloro-o-fluoro, o-fluorobenzylchloride | |
FC1=CC=CC=C1CCl | |
144.57 | |
144.58 |
345-35-7 | |
175°C | |
Pungent | |
1.5154 | |
100 g | |
471699 | |
MOBRMRJUKNQBMY-UHFFFAOYSA-N | |
1-(chloromethyl)-2-fluorobenzene | |
9580 | |
≥98% |
Chemical Identifiers
345-35-7 | |
144.57 | |
MOBRMRJUKNQBMY-UHFFFAOYSA-N | |
9580 | |
FC1=CC=CC=C1CCl |
C7H6ClF | |
MFCD00000892 | |
2-fluorobenzyl chloride, 1-chloromethyl-2-fluorobenzene, o-fluorobenzyl chloride, alpha-chloro-o-fluorotoluene, benzene, 1-chloromethyl-2-fluoro, alpha-chloro-2-fluorotoluene, 2-fluorobenzylchloride, 1-chloromethyl-2-fluoro-benzene, toluene, alpha-chloro-o-fluoro, o-fluorobenzylchloride | |
1-(chloromethyl)-2-fluorobenzene |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 206-460-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only