Please login to your online account to display your discounted pricing

2-Fluorobenzyl bromide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluorobenzyl bromide
Name Note 98%
CAS 446-48-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6BrF
Linear Formula FC6H4CH2Br
Formula Weight 189.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC197960250

  SDS
thermo fisher scientific
197960250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.03
Assay ≥97.5%
Physical Form Liquid
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 05, II, 238
TSCA TSCA
EINECS Number 207-169-1
Density 1.567g/mL
CAS Max % 100.0
Refractive Index 1.5495 to 1.5515 (20°C, 589nm)
Synonym α-Bromo-o-fluorotoluene
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C7H6BrF
CAS 446-48-0
Chemical Name or Material 2-Fluorobenzyl bromide
Assay Percent Range 98%
Linear Formula FC6H4CH2Br
Name Note 98%
MDL Number MFCD00000324
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.