missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzoyl Chloride, TCI America™

Manufacturer:  TCI America F0195100G

 View more versions of this product

Catalog No. F0195100G



Specifications

Specifications

2-Fluorobenzoyl Chloride
Yellow
393-52-2
MFCD00000656
o-fluorobenzoyl chloride, benzoyl chloride, 2-fluoro, benzoyl chloride, o-fluoro, 2-fluorobenzoylchloride, 2-fluoro benzoyl chloride, unii-z4h017n76s, ccris 5914, fluorobenzoyl chloride, benzoyl chloride, fluoro, 2-fluorobenzoic acid chloride
C1=CC=C(C(=C1)C(=O)Cl)F
158.556
158.56
≥98.0% (GC,T)
189°C
100g
C7H4ClFO
3265
RAAGZOYMEQDCTD-UHFFFAOYSA-N
2-fluorobenzoyl chloride
9808
Clear Liquid at 20°C

Chemical Identifiers

393-52-2
158.556
RAAGZOYMEQDCTD-UHFFFAOYSA-N
9808
C1=CC=C(C(=C1)C(=O)Cl)F
C7H4ClFO
MFCD00000656
o-fluorobenzoyl chloride, benzoyl chloride, 2-fluoro, benzoyl chloride, o-fluoro, 2-fluorobenzoylchloride, 2-fluoro benzoyl chloride, unii-z4h017n76s, ccris 5914, fluorobenzoyl chloride, benzoyl chloride, fluoro, 2-fluorobenzoic acid chloride
2-fluorobenzoyl chloride
Safety and Handling

Safety and Handling

RTECSNumber : DM6640000

TSCA : Yes

Documents