missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzoyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC153095000

 View more versions of this product

Catalog No. AC153095000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Fluorobenzoyl chloride
82°C
Glass bottle
1.32
Colorless
500mL
96.0
96% min. (GC)
FC6H4COCl
09, I, 136
Solubility in water: decomposes
C1=CC=C(C(=C1)C(=O)Cl)F
158.56
158.56
97%
1.3200g/mL
Authentic
1.5355 to 1.5375
90.0°C to 92.0°C (15.0mmHg)
4.0°C
393-52-2
100.0
C7H4ClFO
MFCD00000656
2-fluoro benzoyl chloride
RAAGZOYMEQDCTD-UHFFFAOYSA-N
2-fluorobenzoyl chloride
9808
Liquid

Chemical Identifiers

393-52-2
158.56
RAAGZOYMEQDCTD-UHFFFAOYSA-N
9808
C1=CC=C(C(=C1)C(=O)Cl)F
C7H4ClFO
MFCD00000656
2-fluoro benzoyl chloride
2-fluorobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-887-2

SDS Documents