missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzotrichloride, TCI America™

Manufacturer:  TCI America F029025G

 View more versions of this product

Catalog No. F029025G



Specifications

Specifications

2-Fluorobenzotrichloride
Yellow
488-98-2
MFCD00000787
JYLVSNNSOWDQQX-UHFFFAOYSA-N
1-fluoro-2-(trichloromethyl)benzene
68104
Clear Liquid at 20°C
75°C
25g
C7H4Cl3F
2-Fluoro-alpha,alpha,alpha-trichlorotoluene
C1=CC=C(C(=C1)C(Cl)(Cl)Cl)F
213.457
213.46
≥98.0% (GC)

Chemical Identifiers

488-98-2
213.457
JYLVSNNSOWDQQX-UHFFFAOYSA-N
68104
C1=CC=C(C(=C1)C(Cl)(Cl)Cl)F
C7H4Cl3F
MFCD00000787
2-Fluoro-alpha,alpha,alpha-trichlorotoluene
1-fluoro-2-(trichloromethyl)benzene
Safety and Handling

Safety and Handling

TSCA : Yes

Documents