missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluorobenzonitrile, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC250051000
Specifications
2-Fluorobenzonitrile | |
73°C | |
Glass bottle | |
1.11 | |
Brown to Colorless | |
394-47-8 | |
100.0 | |
C7H4FN | |
1-cyano-2-fluorobenzene, 2-cyanofluorobenzene, 2-fluoro-benzonitrile, 2-fluorobenzenecarbonitrile, 2-fluorobenzonitril, benzonitrile, 2-fluoro, benzonitrile, o-fluoro, fluorobenzonitrile, o-cyanofluorobenzene, o-fluorobenzonitrile | |
GDHXJNRAJRCGMX-UHFFFAOYSA-N | |
2-fluorobenzonitrile | |
67855 | |
Liquid |
1.1100g/mL | |
Authentic | |
1.5063 to 1.5083 | |
90°C (21.0mmHg) | |
100g | |
98.5 | |
99% | |
MFCD00001773 | |
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents | |
C1=CC=C(C(=C1)C#N)F | |
121.12 | |
121.12 | |
99% |
Chemical Identifiers
394-47-8 | |
121.12 | |
GDHXJNRAJRCGMX-UHFFFAOYSA-N | |
67855 | |
C1=CC=C(C(=C1)C#N)F |
C7H4FN | |
MFCD00001773 | |
1-cyano-2-fluorobenzene, 2-cyanofluorobenzene, 2-fluoro-benzonitrile, 2-fluorobenzenecarbonitrile, 2-fluorobenzonitril, benzonitrile, 2-fluoro, benzonitrile, o-fluoro, fluorobenzonitrile, o-cyanofluorobenzene, o-fluorobenzonitrile | |
2-fluorobenzonitrile |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre
GHS Signal Word: Warning
EINECSNumber : 206-897-7