missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzoic acid, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 119360250

 View more versions of this product

Catalog No. AC119360250



Description

Description

Specifications

Specifications

2-Fluorobenzoic acid
Plastic bottle
123°C to 125°C
99%
98.5
98.5% min. (GC)
FC6H4CO2H
09, 333
Solubility in water: 7.2g/L (25°C)
C1=CC=C(C(=C1)C(=O)O)F
140.11
CHEBI:19577
Crystalline Powder or Needles
Authentic
White to Yellow
25g
445-29-4
100.0
C7H5FO2
MFCD00002405
2-fluoro-benzoic acid, benzoic acid, 2-fluoro, benzoic acid, o-fluoro, fluorobenzoic acid, o-fluorbenzoesaeure, o-fluorbenzoesaeure german, o-fluorobenzoic acid, ofba, ortho-fluorobenzoic acid, unii-64uz32koo4
NSTREUWFTAOOKS-UHFFFAOYSA-N
2-fluorobenzoic acid
9935
140.11
99%

Chemical Identifiers

445-29-4
140.11
NSTREUWFTAOOKS-UHFFFAOYSA-N
9935
2-fluorobenzoic acid
C7H5FO2
MFCD00002405
2-fluoro-benzoic acid, benzoic acid, 2-fluoro, benzoic acid, o-fluoro, fluorobenzoic acid, o-fluorbenzoesaeure, o-fluorbenzoesaeure german, o-fluorobenzoic acid, ofba, ortho-fluorobenzoic acid, unii-64uz32koo4
CHEBI:19577
C1=CC=C(C(=C1)C(=O)O)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-158-1

RTECSNumber : DH0250000

TSCA : TSCA

SDS Documents