missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzohydrazide 98.0+%, TCI America™

$108.64 - $447.02

Chemical Identifiers

CAS 446-24-2
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
MDL Number MFCD00025112
InChI Key YJCCKQQVXNNAAR-UHFFFAOYSA-N
Synonym 2-fluoranylbenzohydrazide, 2-fluoro-benzohydrazide, 2-fluorobenzenecarbohydrazide, 2-fluorobenzhydrazide, 2-fluorobenzoic acid hydrazide, 2-fluorobenzoic hydrazide, 2-fluorobenzoylhydrazine, acmc-209jyw, o-fluorobenzhydrazide, pubchem3481
PubChem CID 136288
IUPAC Name 2-fluorobenzohydrazide
SMILES NNC(=O)C1=CC=CC=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F06165G
View Documents
Tci America
F06165G
5g
Each for $108.64
 
F061625G
View Documents
Tci America
F061625G
25g
Each for $447.02
 
Description

Description

Specifications

Chemical Identifiers

446-24-2
154.14
YJCCKQQVXNNAAR-UHFFFAOYSA-N
136288
NNC(=O)C1=CC=CC=C1F
C7H7FN2O
MFCD00025112
2-fluoranylbenzohydrazide, 2-fluoro-benzohydrazide, 2-fluorobenzenecarbohydrazide, 2-fluorobenzhydrazide, 2-fluorobenzoic acid hydrazide, 2-fluorobenzoic hydrazide, 2-fluorobenzoylhydrazine, acmc-209jyw, o-fluorobenzhydrazide, pubchem3481
2-fluorobenzohydrazide

Specifications

Undesignated
5g
C7H7FN2O
2-fluoranylbenzohydrazide, 2-fluoro-benzohydrazide, 2-fluorobenzenecarbohydrazide, 2-fluorobenzhydrazide, 2-fluorobenzoic acid hydrazide, 2-fluorobenzoic hydrazide, 2-fluorobenzoylhydrazine, acmc-209jyw, o-fluorobenzhydrazide, pubchem3481
NNC(=O)C1=CC=CC=C1F
154.14
154.14
≥98.0% (GC,T)
73°C
446-24-2
MFCD00025112
YJCCKQQVXNNAAR-UHFFFAOYSA-N
2-fluorobenzohydrazide
136288
Crystal-Powder at 20°C
2-Fluorobenzohydrazide
Safety and Handling

Safety and Handling

TSCA : No

SDS
Product Certifications