missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzamide 98%, ACROS Organics™

$25.16 - $25.16

Chemical Identifiers

CAS 445-28-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H6FNO
Linear Formula FC6H4CONH2
Formula Weight 139.13
MDL Number MFCD00007970
Chemical Name or Material 2-Fluorobenzamide, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC153290050 SDS Thermo Fisher Scientific
153290050
Glass bottle 5g Each for $25.16
Specifications Description & Specifications

Chemical Identifiers

445-28-3
100.0
C7H6FNO
139.13
2-Fluorobenzamide, 98%
97.5
98%
FC6H4CONH2
MFCD00007970
≥97.5%

Specifications

White
5g
Authentic
100.0
Crystalline Powder
C7H6FNO
139.13
2-Fluorobenzamide, 98%
115° to 119°C
445-28-3
97.5
Glass bottle
98%
FC6H4CONH2
MFCD00007970
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-157-6

RTECSNumber : CV4953333

TSCA : TSCA