missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzaldehyde, TCI America™

Manufacturer:  TCI America F0190250ML

 View more versions of this product

Catalog No. F0190250ML



Specifications

Specifications

2-Fluorobenzaldehyde
Yellow
250mL
C7H5FO
1989
ZWDVQMVZZYIAHO-UHFFFAOYSA-N
2-fluorobenzaldehyde
67970
Clear Liquid at 20°C
175°C
-45°C
446-52-6
MFCD00003302
o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
C1=CC=C(C(=C1)C=O)F
124.114
124.11
≥98.0% (GC)

Chemical Identifiers

446-52-6
124.114
ZWDVQMVZZYIAHO-UHFFFAOYSA-N
67970
C1=CC=C(C(=C1)C=O)F
C7H5FO
MFCD00003302
o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
2-fluorobenzaldehyde
Safety and Handling

Safety and Handling

RTECSNumber : CU6140000

TSCA : No

Documents