missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorobenzaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119320250

 View more versions of this product

Catalog No. AC119320250



Description

Description

Specifications

Specifications

2-Fluorobenzaldehyde
55°C
Glass bottle
1.17
Brown to Colorless
25mL
446-52-6
100.0
C7H5FO
MFCD00003302
2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, 7zx20dxb9z, benzaldehyde, 2-fluoro, benzaldehyde, fluoro, benzaldehyde, o-fluoro, fluorobenzaldehyde, o-fluorobenzaldehyde, ortho-fluorobenzaldehyde, unii-7zx20dxb9z
ZWDVQMVZZYIAHO-UHFFFAOYSA-N
2-fluorobenzaldehyde
67970
Liquid
1.1700g/mL
Authentic
1.5205 to 1.5225
172°C to 174°C
-44.5°C
97%
96.0
96% min. (GC)
FC6H4CHO
07, I, 132
Solubility in water: insoluble
C1=CC=C(C(=C1)C=O)F
124.11
124.11
97%

Chemical Identifiers

446-52-6
124.11
ZWDVQMVZZYIAHO-UHFFFAOYSA-N
67970
C1=CC=C(C(=C1)C=O)F
C7H5FO
MFCD00003302
2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, 7zx20dxb9z, benzaldehyde, 2-fluoro, benzaldehyde, fluoro, benzaldehyde, o-fluoro, fluorobenzaldehyde, o-fluorobenzaldehyde, ortho-fluorobenzaldehyde, unii-7zx20dxb9z
2-fluorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-171-2

RTECSNumber : CU6140000

TSCA : TSCA

SDS Documents