Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorobenzal chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluorobenzal chloride
CAS 320-65-0
Molecular Formula C7H5Cl2F
Formula Weight 179.02
MDL Number MFCD00043797
UN Number UN3265
Beilstein 2042648
Synonym alpha,alpha-Dichloro-2-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0398406

  SDS
alfa aesar™
L0398406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0398414

  SDS
alfa aesar™
L0398414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0398422

  SDS
alfa aesar™
L0398422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.02
Chemical Name or Material 2-Fluorobenzal chloride
CAS 320-65-0
Beilstein 2042648
MDL Number MFCD00043797
Assay 97%
Boiling Point 224° to 232°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Quantity 5g
Synonym alpha,alpha-Dichloro-2-fluorotoluene
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
UN Number UN3265
Molecular Formula C7H5Cl2F
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.