Please login to your online account to display your discounted pricing

2-Fluoroanisole 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoroanisole
Name Note 98%
CAS 321-28-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C7H7FO
Linear Formula FC6H4OCH3
Formula Weight 126.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119301000

  SDS
thermo fisher scientific
119301000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Crystalline Powder
Formula Weight 126.13
Quantity 100mL
Color Orange
TSCA TSCA
EINECS Number 206-284-4
Density 1.1200g/mL
Beilstein 06, II, 169
CAS Max % 100.0
Refractive Index 1.4940 to 1.4970 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Assay Percent Range 98%
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00000316
CAS 321-28-8
Molecular Formula C7H7FO
Chemical Name or Material 2-Fluoroanisole
Linear Formula FC6H4OCH3
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.