Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoroadenosine, 97%

$235.51 - $711.15

Chemical Identifiers

Chemical Name or Material 2-Fluoroadenosine
CAS 146-78-1
Molecular Formula C10H12FN5O4
Formula Weight 285.23
MDL Number MFCD00866394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741203

 
alfa aesar™
H2741203
1g Each for $711.15

AAH27412MD

 
alfa aesar™
H27412MD
250mg Each for $235.51
Description & Specifications

Specifications

Formula Weight 285.23
Chemical Name or Material 2-Fluoroadenosine
Assay 97%
Molecular Formula C10H12FN5O4
MDL Number MFCD00866394
CAS 146-78-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number AU7386000
Optical Rotation −72° (c=0.1 in EtOH)
Melting Point 231°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.

2° to 8°C