missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2′-Fluoroacetophenone, TCI America™

Manufacturer:  TCI America F036425G

 View more versions of this product

Catalog No. F036425G



Specifications

Specifications

2′-Fluoroacetophenone
Yellow
445-27-2
MFCD00000320
QMATYTFXDIWACW-UHFFFAOYSA-N
1-(2-fluorophenyl)ethanone
96744
Clear Liquid at 20°C
85°C
25g
C8H7FO
2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2-fluoroacetophenone, 2;-fluoroacetophenone
CC(=O)C1=CC=CC=C1F
138.141
138.14
≥97.0% (GC)

Chemical Identifiers

445-27-2
138.141
QMATYTFXDIWACW-UHFFFAOYSA-N
96744
CC(=O)C1=CC=CC=C1F
C8H7FO
MFCD00000320
2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2-fluoroacetophenone, 2;-fluoroacetophenone
1-(2-fluorophenyl)ethanone
Safety and Handling

Safety and Handling

TSCA : No

Documents