Please login to your online account to display your discounted pricing

2'-Fluoroacetophenone, 97%, ACROS Organics™

$226.17 - $226.17

Chemical Identifiers

Chemical Name or Material 2';-Fluoroacetophenone
Name Note 97%
CAS 445-27-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7FO
Linear Formula FC6H4COCH3
Formula Weight 138.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC179780250

  SDS
thermo fisher scientific
179780250
Glass bottle 25g Each for $226.17
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 138.14
Quantity 25g
Color Clear colorless to yellow
EINECS Number 207-156-0
TSCA TSCA
Density 1.1370g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5065 to 1.5085 (20°C, 589nm)
Synonym 1-(2-Fluorophenyl)ethanone
Infrared Spectrum Authentic
Chemical Name or Material 2';-Fluoroacetophenone
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H7FO
Linear Formula FC6H4COCH3
MDL Number MFCD00000320
CAS 445-27-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.