Please login to your online account to display your discounted pricing

2'-Fluoroacetanilide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  305840050

 View more versions of this product

Catalog No. AC305840050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 153.16
Quantity 5g
Color White
EINECS Number 206-916-9
TSCA TSCA
Melting Point 76° to 80°C
CAS Max % 100.0
Chemical Name or Material 2'-Fluoroacetanilide
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H8FNO
MDL Number MFCD00000315
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula CH3CONHC6H4F
CAS 399-31-5

Specifications

Chemical Name or Material 2'-Fluoroacetanilide
Name Note 98%
CAS 399-31-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8FNO
Linear Formula CH3CONHC6H4F
Formula Weight 153.16
MDL Number MFCD00000315
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents