Please login to your online account to display your discounted pricing

2'-Fluoroacetanilide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2'-Fluoroacetanilide
Name Note 98%
CAS 399-31-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8FNO
Linear Formula CH3CONHC6H4F
Formula Weight 153.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305840050

  SDS
thermo fisher scientific
305840050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 153.16
Quantity 5g
Color White
EINECS Number 206-916-9
TSCA TSCA
Melting Point 76° to 80°C
CAS Max % 100.0
Chemical Name or Material 2'-Fluoroacetanilide
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H8FNO
MDL Number MFCD00000315
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula CH3CONHC6H4F
CAS 399-31-5
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.