Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Fluoroacetanilide, 98+%

$54.65 - $169.32

Chemical Identifiers

Chemical Name or Material 2'-Fluoroacetanilide
CAS 399-31-5
Molecular Formula C8H8FNO
Formula Weight 153.16
MDL Number MFCD00000315
Beilstein 2717443
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1692714

  SDS
alfa aesar™
A1692714
25g Each for $54.65

AAA1692722

  SDS
alfa aesar™
A1692722
100g Each for $169.32
Description & Specifications

Specifications

Formula Weight 153.16
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2'-Fluoroacetanilide
Assay ≥98%
Molecular Formula C8H8FNO
MDL Number MFCD00000315
CAS 399-31-5
Beilstein 2717443
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 140° to 142°C (14mmHg)
Melting Point 76° to 80°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.