Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-N-phenylbenzamide, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-N-phenylbenzamide
CAS 1747-80-4
Formula Weight 215.23
MDL Number MFCD00454563
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5906303

 
alfa aesar™
H5906303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH59063MD

 
alfa aesar™
H59063MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.23
Chemical Name or Material 2-Fluoro-N-phenylbenzamide
Physical Form Solid
Assay 97%
MDL Number MFCD00454563
CAS 1747-80-4
Quantity 1g