missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-N,N-dimethyl-4-nitroaniline, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 65739-04-0
Molecular Formula C8H9FN2O2
Molecular Weight (g/mol) 184.17
InChI Key QTXUWCWOGLFTTP-UHFFFAOYSA-N
Synonym 2-fluoro-n,n-dimethyl-4-nitro-benzenamine, acmc-209nru, benzenamine, 2-fluoro-n,n-dimethyl-4-nitro, n,n-dimethyl-2-fluoro-4-nitroaniline
PubChem CID 23547995
IUPAC Name 2-fluoro-N,N-dimethyl-4-nitroaniline
SMILES CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63930MD View Documents Alfa Aesar™
H63930MD
250mg Each for $28.84
 
AAH6393003 View Documents Alfa Aesar™
H6393003
1g Each for $85.14
 
AAH6393006 View Documents Alfa Aesar™
H6393006
5g Each for $337.73
 
Specifications

Chemical Identifiers

65739-04-0
184.17
2-fluoro-n,n-dimethyl-4-nitro-benzenamine, acmc-209nru, benzenamine, 2-fluoro-n,n-dimethyl-4-nitro, n,n-dimethyl-2-fluoro-4-nitroaniline
2-fluoro-N,N-dimethyl-4-nitroaniline
C8H9FN2O2
QTXUWCWOGLFTTP-UHFFFAOYSA-N
23547995
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

250mg
C8H9FN2O2
2-fluoro-n,n-dimethyl-4-nitro-benzenamine, acmc-209nru, benzenamine, 2-fluoro-n,n-dimethyl-4-nitro, n,n-dimethyl-2-fluoro-4-nitroaniline
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])F
184.17
184.17
97%
65739-04-0
UN2811
QTXUWCWOGLFTTP-UHFFFAOYSA-N
2-fluoro-N,N-dimethyl-4-nitroaniline
23547995
Solid
2-Fluoro-N,N-dimethyl-4-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.