missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-N-methylbenzylamine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5443906
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and air. Store at 4-2°C.
Specifications
2-Fluoro-N-methylbenzylamine | |
Colorless to Tan | |
73°C | |
C8H10FN | |
MFCD04629633 | |
Air Sensitive | |
Slightly soluble in water. | |
CNCC1=CC=CC=C1F | |
139.173 | |
139.17 | |
Liquid |
399-30-4 | |
1.047 g/mL | |
68°C (154°F) | |
1.047 | |
5 g | |
n-methyl-2-fluorobenzylamine, 2-fluorobenzyl methylamine, 2-fluoro-n-methylbenzylamine, 2-fluorophenyl methyl methyl amine, 1-2-fluorophenyl-n-methylmethanamine, n-2-fluorobenzyl-n-methylamine, n-methyl-o-fluorobenzylamine, benzenemethanamine, 2-fluoro-n-methyl, 1-2-fluorophenyl-n-methyl-methanamine, acmc-209zio | |
AHIHZCXUWGORQO-UHFFFAOYSA-N | |
1-(2-fluorophenyl)-N-methylmethanamine | |
2060842 | |
95% |
Chemical Identifiers
399-30-4 | |
139.173 | |
AHIHZCXUWGORQO-UHFFFAOYSA-N | |
2060842 | |
CNCC1=CC=CC=C1F |
C8H10FN | |
MFCD04629633 | |
n-methyl-2-fluorobenzylamine, 2-fluorobenzyl methylamine, 2-fluoro-n-methylbenzylamine, 2-fluorophenyl methyl methyl amine, 1-2-fluorophenyl-n-methylmethanamine, n-2-fluorobenzyl-n-methylamine, n-methyl-o-fluorobenzylamine, benzenemethanamine, 2-fluoro-n-methyl, 1-2-fluorophenyl-n-methyl-methanamine, acmc-209zio | |
1-(2-fluorophenyl)-N-methylmethanamine |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only