missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-N-methylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H56104MD

 View more versions of this product

Catalog No. AAH56104MD



Specifications

Specifications

2-Fluoro-N-methylbenzamide
52833-63-3
MFCD01214098
NAGFMACWWJYORB-UHFFFAOYSA-N
2-fluoro-N-methylbenzamide
2425972
97%
250mg
C8H8FNO
2-fluoro-n-methyl-benzamide
CNC(=O)C1=CC=CC=C1F
153.156
153.16

Chemical Identifiers

52833-63-3
153.156
NAGFMACWWJYORB-UHFFFAOYSA-N
2425972
CNC(=O)C1=CC=CC=C1F
C8H8FNO
MFCD01214098
2-fluoro-n-methyl-benzamide
2-fluoro-N-methylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents