Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-N-methyl-6-phenoxybenzylamine hydrochloride, 98%

$155.04 - $155.04

Chemical Identifiers

Chemical Name or Material 2-Fluoro-N-methyl-6-phenoxybenzylamine hydrochloride
CAS 902836-71-9
Molecular Formula C14H14FNO·HCl
Formula Weight 267.73g/mol
MDL Number MFCD12546206
Synonym 1-(2-Fluoro-6-phenoxyphenyl)-N-methylmethylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5168803

 
alfa aesar™
H51688-03
Each for $155.04
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2-Fluoro-N-methyl-6-phenoxybenzylamine hydrochloride
CAS 902836-71-9
Assay 98%
Molecular Formula C14H14FNO·HCl
Formula Weight 267.73g/mol
MDL Number MFCD12546206
Synonym 1-(2-Fluoro-6-phenoxyphenyl)-N-methylmethylamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.