missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-N-(3,5-dimethylphenyl)benzamide, 97%

Manufacturer:  Alfa Aesar™ H5970503

 View more versions of this product

Catalog No. AAH5970503



Specifications

Specifications

2-Fluoro-N-(3,5-dimethylphenyl)benzamide
332168-85-1
MFCD01115438
GKEBWXLAFINVJG-UHFFFAOYSA-N
N-(3,5-dimethylphenyl)-2-fluorobenzamide
803512
Solid
1g
C15H14FNO
2-fluoro-n-3,5-dimethylphenyl benzamide
CC1=CC(=CC(=C1)NC(=O)C2=CC=CC=C2F)C
243.281
243.28
97%

Chemical Identifiers

332168-85-1
243.281
GKEBWXLAFINVJG-UHFFFAOYSA-N
803512
CC1=CC(=CC(=C1)NC(=O)C2=CC=CC=C2F)C
C15H14FNO
MFCD01115438
2-fluoro-n-3,5-dimethylphenyl benzamide
N-(3,5-dimethylphenyl)-2-fluorobenzamide
Documents