Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-N-(3,5-dimethylphenyl)benzamide, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-N-(3,5-dimethylphenyl)benzamide
CAS 332168-85-1
Formula Weight 243.28
MDL Number MFCD01115438
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5970503

 
alfa aesar™
H5970503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH59705MD

 
alfa aesar™
H59705MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.28
MDL Number MFCD01115438
CAS 332168-85-1
Quantity 1g
Physical Form Solid
Chemical Name or Material 2-Fluoro-N-(3,5-dimethylphenyl)benzamide
Assay 97%