Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-L-phenylalanine, 99+% (sum of enantiomers)

$90.05 - $226.27

Chemical Identifiers

Chemical Name or Material 2-Fluoro-L-phenylalanine
CAS 19883-78-4
Name Note sum of enantiomers
Molecular Formula C9H10FNO2
Formula Weight 183.18
MDL Number MFCD00066448
Synonym H-Phe(2-F)-OH; o-Fluoro-L-phenylalanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2780703

  SDS
alfa aesar™
H2780703
1g Each for $226.27

AAH27807MD

  SDS
alfa aesar™
H27807MD
250mg Each for $90.05
Description & Specifications

Specifications

Formula Weight 183.18
Chemical Name or Material 2-Fluoro-L-phenylalanine
Name Note sum of enantiomers
Assay ≥99%
Molecular Formula C9H10FNO2
MDL Number MFCD00066448
CAS 19883-78-4
Synonym H-Phe(2-F)-OH; o-Fluoro-L-phenylalanine
Quantity 1g