missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-DL-phenylalanine, TCI America™

Manufacturer:  TCI America F01701G

 View more versions of this product

Catalog No. F01701G



Specifications

Specifications

2-Fluoro-DL-phenylalanine
239°C
2629-55-2
MFCD00004271
NYCRCTMDYITATC-UHFFFAOYSA-N
2-amino-3-(2-fluorophenyl)propanoic acid
9465
Crystal-Powder at 20°C
White-Yellow
1g
C9H10FNO2
2-fluoro-dl-phenylalanine, 2-amino-3-2-fluorophenyl propanoic acid, 2-fluorophenylalanine, o-fluoro-dl-phenylalanine, o-fluorophenylalanine, 3-o-fluorophenyl alanine, dl-2-fluorophenyl alanine, dl-2-f-phe-oh, dl-o-fluorophenylalanine, dl-beta-o-fluorophenylalanine
C1=CC=C(C(=C1)CC(C(=O)O)N)F
183.182
183.18
≥97.0% (T)

Chemical Identifiers

2629-55-2
183.182
NYCRCTMDYITATC-UHFFFAOYSA-N
9465
C1=CC=C(C(=C1)CC(C(=O)O)N)F
C9H10FNO2
MFCD00004271
2-fluoro-dl-phenylalanine, 2-amino-3-2-fluorophenyl propanoic acid, 2-fluorophenylalanine, o-fluoro-dl-phenylalanine, o-fluorophenylalanine, 3-o-fluorophenyl alanine, dl-2-fluorophenyl alanine, dl-2-f-phe-oh, dl-o-fluorophenylalanine, dl-beta-o-fluorophenylalanine
2-amino-3-(2-fluorophenyl)propanoic acid
Safety and Handling

Safety and Handling

RTECSNumber : AY6307000

TSCA : No

Documents