Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-D-phenylalanine, 99+% (sum of enantiomers)

$70.19 - $190.86

Chemical Identifiers

Chemical Name or Material 2-Fluoro-D-phenylalanine
CAS 97731-02-7
Name Note sum of enantiomers
Molecular Formula C9H10FNO2
Formula Weight 183.18
MDL Number MFCD00066447
Synonym H-D-Phe(2-F)-OH; o-Fluoro-D-phenylalanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2752403

 
alfa aesar™
H2752403
1g Each for $190.86

AAH27524MD

 
alfa aesar™
H27524MD
250mg Each for $70.19
Description & Specifications

Specifications

Formula Weight 183.18
Chemical Name or Material 2-Fluoro-D-phenylalanine
Name Note sum of enantiomers
Assay ≥99%
Molecular Formula C9H10FNO2
MDL Number MFCD00066447
CAS 97731-02-7
Synonym H-D-Phe(2-F)-OH; o-Fluoro-D-phenylalanine
Melting Point 209° to 211°C
Quantity 1g