Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)pyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)pyridine
CAS 94239-04-0
Molecular Formula C6H3F4N
Formula Weight 165.09
MDL Number MFCD03092901
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3263303

  SDS
alfa aesar™
H3263303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3263306

  SDS
alfa aesar™
H3263306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.09
Health Hazard 2 GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Quantity 1g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Molecular Formula C6H3F4N
UN Number UN1993
Health Hazard 1 Warning
Assay 98%
MDL Number MFCD03092901
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)pyridine
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 94239-04-0
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.